siRNA Details
| Gene Symbol: RBM41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171080.1 | NP_001164551.1 | 5 | 865 | ||
| NM_001324242.1 | NP_001311171.1 | 6 | 937 | |||
| NM_001324243.1 | NP_001311172.1 | 6 | 1030 | |||
| NM_001324244.1 | NP_001311173.1 | 6 | 937 | |||
| NM_018301.4 | NP_060771.3 | 5 | 865 | |||
| XM_011530980.2 | XP_011529282.1 | 6 | 1029 | |||
| XM_011530981.2 | XP_011529283.1 | 5 | 957 | |||
| GenBank mRNA | AK001878.1 | - | 5 | 838 | ||
| BC006986.1 | - | 5 | 842 | 3995903 | ||

RBM41
Genomic Map