siRNA Details
| Gene Symbol: C4orf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104629.1 | NP_001098099.1 | 4 | 935 | ||
| NM_018302.2 | NP_060772.2 | 3 | 930 | |||
| XM_011513712.2 | XP_011512014.1 | 5 | 999 | |||
| XM_011513713.2 | XP_011512015.1 | 4 | 1201 | |||
| XM_017008361.1 | XP_016863850.1 | 2 | 2131 | |||
| GenBank mRNA | AK001879.1 | - | 3 | 930 | ||
| AK225192.1 | - | 3 | 899 | |||
| BC037906.1 | - | 4 | 896 | 5276648 | ||

C4orf19
Genomic Map