siRNA Details
| Gene Symbol: UFSP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018359.3 | NP_060829.2 | 11 | 1365 | ||
| NR_028085.1 | 11 | 1340 | ||||
| GenBank mRNA | AK002062.1 | - | 11 | 1294 | ||
| AK091635.1 | - | 11 | 1333 | |||
| BC010493.1 | - | 10 | 1261 | 3048997 | ||
| CA432183.1 | - | 2 | 591 | |||
| CR457278.1 | - | 10 | 1248 | |||

UFSP2
Genomic Map