siRNA Details
| Gene Symbol: DRAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018370.2 | NP_060840.2 | 4 | 858 | ||
| XM_005269004.2 | XP_005269061.1 | 4 | 743 | |||
| XM_017019578.1 | XP_016875067.1 | 5 | 980 | |||
| GenBank mRNA | AK002121.1 | - | 2 | 164 | ||
| AK094923.1 | - | 4 | 488 | |||
| BC013773.1 | - | 2 | 156 | |||
| BC018435.1 | - | 4 | 834 | 4474297 | ||

DRAM1
Genomic Map