siRNA Details
| Gene Symbol: LAPTM4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018407.4 | NP_060877.3 | 1, 2 | 522 | ||
| GenBank mRNA | AF317417.1 | - | 1, 2 | 93 | ||
| AF527412.1 | - | 1, 2 | 451 | |||
| AK075326.1 | - | 1, 2 | 284 | |||
| AL136942.1 | - | 1, 2 | 276 | |||
| AY057051.2 | - | 1, 2 | 522 | |||
| AY261384.1 | - | 1, 2 | 541 | |||
| BC014129.2 | - | 1, 2 | 278 | 4581284 | ||
| BC031021.1 | - | 1, 2 | 303 | 4690368 | ||
| BC038117.1 | - | 1, 2 | 1070 | 5264567 | ||

LAPTM4B
Genomic Map