siRNA Details
| Gene Symbol: TMEM176A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018487.2 | NP_060957.2 | 7 | 761 | ||
| XM_011516376.2 | XP_011514678.1 | 7 | 896 | |||
| XM_011516377.2 | XP_011514679.1 | 7 | 912 | |||
| XM_011516378.2 | XP_011514680.1 | 6 | 794 | |||
| XM_017012393.1 | XP_016867882.1 | 7 | 997 | |||
| GenBank mRNA | AF258340.1 | - | 7 | 736 | ||
| AK123468.1 | - | 2 | 1787 | |||
| BC008303.1 | - | 7 | 779 | 3536986 | ||
| BM663079.1 | - | 1 | 291 | |||

TMEM176A
Genomic Map