siRNA Details
| Gene Symbol: LRRC59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018509.3 | NP_060979.2 | 4 | 626 | ||
| GenBank mRNA | AK025328.1 | - | 4 | 626 | ||
| AK316593.1 | - | 2 | 128 | |||
| BC017168.1 | - | 4 | 584 | 3907072 | ||
| BC052279.1 | - | 4 | 544 | 4127952 | ||

LRRC59
Genomic Map