siRNA Details
| Gene Symbol: DNAJA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130182.1 | NP_001123654.1 | 3 | 451 | ||
| NM_001130183.1 | NP_001123655.1 | 3 | 278 | |||
| NM_018602.3 | NP_061072.3 | 4 | 590 | |||
| XM_005254535.3 | XP_005254592.1 | 4 | 363 | |||
| XM_006720601.1 | XP_006720664.1 | 4 | 365 | |||
| GenBank mRNA | AF116663.1 | - | 2 | 682 | ||
| AK096616.1 | - | 3 | 451 | |||
| AK098079.1 | - | 4 | 581 | |||
| BC021720.2 | - | 4 | 590 | 4831235 | ||
| BC031044.1 | - | 3 | 313 | 5277834 | ||
| BX648710.1 | - | 4 | 472 | |||
| DA561365.1 | - | 3 | 279 | |||

DNAJA4
Genomic Map