siRNA Details
| Gene Symbol: FEM1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018708.2 | NP_061178.1 | 1 | 2126 | ||
| GenBank mRNA | AJ311359.1 | - | 1 | 1986 | ||
| AK074232.1 | - | 1 | 2084 | |||
| AK126532.1 | - | 2 | 1854 | |||
| AK315554.1 | - | 1 | 2098 | |||
| AL359589.1 | - | 1 | 264 | |||
| AL365408.1 | - | 1 | 581 | |||
| AY356352.1 | - | 1 | 2080 | |||
| BC004988.1 | - | 1 | 2084 | 2957950 | ||
| CR749732.1 | - | 1 | 264 | |||

FEM1A
Genomic Map