siRNA Details
| Gene Symbol: TMEM55A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018710.2 | NP_061180.1 | 7 | 941 | ||
| GenBank mRNA | AK313783.1 | - | 7 | 854 | ||
| AX775961.1 | - | 7 | 791 | |||
| BC033892.1 | - | 7 | 791 | 4820065 | ||
| CR749733.1 | - | 6 | 505 | |||

TMEM55A
Genomic Map