siRNA Details
| Gene Symbol: PLXNA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017514.4 | NP_059984.3 | 24 | 4438 | ||
| XM_005274705.4 | XP_005274762.2 | 24 | 4378 | |||
| XM_005274706.4 | XP_005274763.2 | 24 | 4110 | |||
| XM_006724829.3 | XP_006724892.1 | 24 | 4342 | |||
| XM_011531183.2 | XP_011529485.1 | 23 | 3802 | |||
| GenBank mRNA | X87852.1 | - | 24 | 4447 | ||

PLXNA3
Genomic Map