siRNA Details
| Gene Symbol: PLXNA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017514.4 | NP_059984.3 | 28 | 4910 | ||
| XM_005274705.4 | XP_005274762.2 | 28 | 4850 | |||
| XM_005274706.4 | XP_005274763.2 | 28 | 4582 | |||
| XM_006724829.3 | XP_006724892.1 | 28 | 4814 | |||
| XM_011531183.2 | XP_011529485.1 | 27 | 4274 | |||
| GenBank mRNA | X87852.1 | - | 28 | 4919 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008883, D86950, AK220285, AK049319, BC093482.

PLXNA3
Genomic Map