siRNA Details
| Gene Symbol: UBE2Q1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017582.6 | NP_060052.3 | 2, 3 | 523 | ||
| GenBank mRNA | AK293986.1 | - | 2, 3 | 146 | ||
| AY112698.1 | - | 2, 3 | 498 | |||
| AY948200.1 | - | 2, 3 | 506 | |||
| BC070158.1 | - | 1, 2 | 48 | |||
| BC107057.2 | - | 3, 4 | 191 | |||
| BC148185.2 | - | 2, 3 | 200 | |||
| BQ278712.1 | - | 523 | ||||

UBE2Q1
Genomic Map