siRNA Details
| Gene Symbol: DDX60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017631.5 | NP_060101.3 | 15 | 2362 | ||
| XM_011532103.2 | XP_011530405.1 | 15 | 2362 | |||
| XM_011532104.2 | XP_011530406.1 | 15 | 2362 | |||
| XM_017008383.1 | XP_016863872.1 | 15 | 2362 | |||
| XM_017008384.1 | XP_016863873.1 | 15 | 2362 | |||
| GenBank mRNA | AK001649.1 | - | 15 | 2238 | ||
| AK307404.1 | - | 15 | 2362 | |||
| BC038115.2 | - | 15 | 2307 | 5261001 | ||

DDX60
Genomic Map