siRNA Details
| Gene Symbol: ANKRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017664.3 | NP_060134.2 | 6 | 1233 | ||
| NR_104587.1 | 7 | 1295 | ||||
| GenBank mRNA | AK000100.1 | - | 6 | 1195 | ||
| AK000111.1 | - | 1 | 526 | |||
| AK026017.1 | - | 2 | 307 | |||
| AK095763.1 | - | 4 | 905 | |||
| AK096083.1 | - | 3 | 603 | |||
| AK123341.1 | - | 1 | 1068 | |||
| AK125100.1 | - | 1 | 2201 | |||
| BX641022.1 | - | 1 | 2229 | |||

ANKRD10
Genomic Map