siRNA Details
| Gene Symbol: ANKRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286721.1 | NP_001273650.1 | 1, 2 | 381 | ||
| NM_017664.3 | NP_060134.2 | 1, 2 | 381 | |||
| NR_104586.1 | 1, 2 | 381 | ||||
| NR_104587.1 | 1, 2 | 381 | ||||
| GenBank mRNA | AK000100.1 | - | 1, 2 | 343 | ||
| AK096083.1 | - | 1, 2 | 301 | |||
| BC001727.1 | - | 1, 2 | 280 | 3354202 | ||
| BC039715.1 | - | 1, 2 | 311 | |||
| BX354283.2 | - | 1, 2 | 277 | |||
| DA961131.1 | - | 1, 2 | 381 | |||

ANKRD10
Genomic Map