siRNA Details
| Gene Symbol: FERMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017671.4 | NP_060141.3 | 5 | 1503 | ||
| GenBank mRNA | AB105105.1 | - | 5 | 1503 | ||
| AF443278.1 | - | 5 | 1072 | |||
| AK092195.1 | - | 3 | 561 | |||
| AK314394.1 | - | 5 | 893 | |||
| AY137240.1 | - | 4 | 714 | |||
| BC035882.1 | - | 5 | 1186 | 4941567 | ||

FERMT1
Genomic Map