siRNA Details
| Gene Symbol: FERMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017671.4 | NP_060141.3 | 6, 7 | 1643 | ||
| GenBank mRNA | AB105105.1 | - | 6, 7 | 1643 | ||
| AF443278.1 | - | 6, 7 | 1212 | |||
| AK025365.1 | - | 1, 2 | 24 | |||
| AK092195.1 | - | 4, 5 | 701 | |||
| AK314394.1 | - | 6, 7 | 1033 | |||
| AY137240.1 | - | 5, 6 | 854 | |||
| BC035882.1 | - | 6, 7 | 1326 | 4941567 | ||
| BX647465.1 | - | 1, 2 | 50 | |||

FERMT1
Genomic Map