siRNA Details
| Gene Symbol: LRRC40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017768.4 | NP_060238.3 | 5 | 745 | ||
| XM_005271013.1 | XP_005271070.1 | 4 | 572 | |||
| XM_011541763.1 | XP_011540065.1 | 3 | 487 | |||
| GenBank mRNA | AK000338.1 | - | 3 | 255 | ||
| AK022951.1 | - | 5 | 659 | |||
| AK225337.1 | - | 3 | 255 | |||
| BC003407.1 | - | 5 | 545 | |||
| BC008586.1 | - | 5 | 643 | 2900193 | ||

LRRC40
Genomic Map