siRNA Details
| Gene Symbol: RAB20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017817.2 | NP_060287.1 | 2 | 798 | ||
| GenBank mRNA | AJ272065.1 | - | 2 | 580 | ||
| AK000436.1 | - | 2 | 785 | |||
| AK000446.1 | - | 2 | 785 | |||
| BC026025.1 | - | 2 | 794 | 4636491 | ||

RAB20
Genomic Map