siRNA Details
| Gene Symbol: TMEM127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193304.2 | NP_001180233.1 | 2, 3 | 497 | ||
| NM_017849.3 | NP_060319.1 | 2, 3 | 520 | |||
| GenBank mRNA | AK000514.1 | - | 2, 3 | 501 | ||
| AK293276.1 | - | 4, 5 | 401 | |||
| BC028575.1 | - | 2, 3 | 478 | 4837629 | ||
| BC039892.1 | - | 2, 3 | 473 | 5725347 | ||

TMEM127
Genomic Map