siRNA Details
| Gene Symbol: ZNF416 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017879.1 | NP_060349.1 | 1, 2 | 193 | ||
| XM_005259051.3 | XP_005259108.1 | 1, 2 | 256 | |||
| GenBank mRNA | AK000564.1 | - | 1, 2 | 245 | ||
| BC000130.1 | - | 1, 2 | 193 | 2901256 | ||

ZNF416
Genomic Map