siRNA Details
| Gene Symbol: NPLOC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017921.3 | NP_060391.2 | 1, 2 | 238 | ||
| XM_011524979.1 | XP_011523281.1 | 1, 2 | 233 | |||
| XM_011524980.1 | XP_011523282.1 | 1, 2 | 233 | |||
| XM_011524981.1 | XP_011523283.1 | 1, 2 | 233 | |||
| XM_011524982.2 | XP_011523284.1 | 1, 2 | 233 | |||
| GenBank mRNA | AB040932.1 | - | 1, 2 | 145 | ||
| AK000664.1 | - | 1, 2 | 147 | |||
| AK294473.1 | - | 1, 2 | 164 | |||
| BC025930.1 | - | 1, 2 | 179 | 5017718 | ||
| DC322260.1 | - | 1, 2 | 197 | |||
| HY116973.1 | - | 1, 2 | 240 | |||

NPLOC4
Genomic Map