siRNA Details
| Gene Symbol: GPATCH2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322028.1 | NP_001308957.1 | 9 | 1379 | ||
| NM_001322029.1 | NP_001308958.1 | 9 | 1379 | |||
| NM_001322030.1 | NP_001308959.1 | 9 | 1379 | |||
| NM_017926.3 | NP_060396.2 | 9 | 1379 | |||
| NM_017972.3 | NP_060442.2 | 8 | 1257 | |||
| NR_110314.1 | 9 | 1372 | ||||
| XM_006720191.2 | XP_006720254.1 | 9 | 1337 | |||
| XM_017021427.1 | XP_016876916.1 | 10 | 1579 | |||
| XM_017021428.1 | XP_016876917.1 | 10 | 1564 | |||
| XM_017021429.1 | XP_016876918.1 | 10 | 1579 | |||
| XM_017021430.1 | XP_016876919.1 | 10 | 1564 | |||
| XM_017021431.1 | XP_016876920.1 | 10 | 1579 | |||
| XM_017021432.1 | XP_016876921.1 | 10 | 1579 | |||
| XM_017021433.1 | XP_016876922.1 | 9 | 1337 | |||
| GenBank mRNA | AK000696.1 | - | 9 | 1316 | ||
| AK000895.1 | - | 8 | 722 | |||
| BC038835.1 | - | 9 | 1068 | |||

GPATCH2L
Genomic Map