siRNA Details
| Gene Symbol: MFN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033540.2 | NP_284941.2 | 6 | 697 | ||
| XM_005247596.3 | XP_005247653.2 | 6 | 655 | |||
| XM_011512963.2 | XP_011511265.1 | 3 | 283 | |||
| GenBank mRNA | AB043588.1 | - | 5 | 662 | ||
| AB043589.1 | - | 6 | 662 | |||
| AF329637.1 | - | 5 | 571 | |||
| AK000700.1 | - | 6 | 654 | |||
| AK314306.1 | - | 6 | 654 | |||
| BC020661.1 | - | 6 | 663 | |||
| BC040557.1 | - | 6 | 697 | 5270347 | ||
| CR627379.1 | - | 3 | 347 | |||
| U95822.1 | - | 2 | 160 | |||

MFN1
Genomic Map