siRNA Details
| Gene Symbol: SCYL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317784.1 | NP_001304713.1 | 3 | 564 | ||
| NM_017988.5 | NP_060458.3 | 3 | 716 | |||
| XM_017019618.1 | XP_016875107.1 | 3 | 564 | |||
| XM_017019619.1 | XP_016875108.1 | 3 | 716 | |||
| XM_017019620.1 | XP_016875109.1 | 3 | 567 | |||
| GenBank mRNA | AK000936.1 | - | 2 | 322 | ||
| AK125091.1 | - | 3 | 701 | |||
| AK292980.1 | - | 3 | 490 | |||
| BC063798.1 | - | 3 | 329 | 6473586 | ||
| DA234206.1 | - | 3 | 564 | |||

SCYL2
Genomic Map