siRNA Details
| Gene Symbol: MREG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018000.2 | NP_060470.2 | 1, 2 | 393 | ||
| XM_011511468.2 | XP_011509770.1 | 1, 2 | 243 | |||
| XM_017004475.1 | XP_016859964.1 | 1, 2 | 243 | |||
| GenBank mRNA | AK000978.1 | - | 1, 2 | 363 | ||
| BC032747.1 | - | 1, 2 | 402 | 5573149 | ||
| BC082990.1 | - | 1, 2 | 383 | 6159624 | ||
| BQ928130.1 | - | 1, 2 | 393 | |||
| CX781801.1 | - | 1, 2 | 388 | |||

MREG
Genomic Map