siRNA Details
| Gene Symbol: ARHGEF40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278529.1 | NP_001265458.1 | 3 | 586 | ||
| NM_001278530.1 | NP_001265459.1 | 3 | 586 | |||
| NM_018071.4 | NP_060541.3 | 3 | 586 | |||
| XM_005267844.2 | XP_005267901.1 | 3 | 622 | |||
| XM_011536937.2 | XP_011535239.1 | 3 | 622 | |||
| XM_011536938.2 | XP_011535240.1 | 3 | 624 | |||
| XM_017021434.1 | XP_016876923.1 | 4 | 879 | |||
| XM_017021435.1 | XP_016876924.1 | 3 | 623 | |||
| GenBank mRNA | AK074057.1 | - | 3 | 531 | ||
| BC142692.1 | - | 3 | 529 | 40148708 | ||
| BC151212.1 | - | 3 | 529 | |||
| BX248768.1 | - | 3 | 573 | |||

ARHGEF40
Genomic Map