siRNA Details
| Gene Symbol: IPO9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018085.4 | NP_060555.2 | 4 | 399 | ||
| GenBank mRNA | AF410465.1 | - | 4 | 349 | ||
| DA861528.1 | - | 4 | 362 | |||
| DB232332.1 | - | 5 | 510 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IPO9| Gene Symbol: IPO9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018085.4 | NP_060555.2 | 4 | 399 | ||
| GenBank mRNA | AF410465.1 | - | 4 | 349 | ||
| DA861528.1 | - | 4 | 362 | |||
| DB232332.1 | - | 5 | 510 | |||