siRNA Details
| Gene Symbol: ASUN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018164.2 | NP_060634.2 | 6 | 1178 | ||
| XM_005253422.3 | XP_005253479.1 | 6 | 1137 | |||
| XM_011520750.2 | XP_011519052.1 | 6 | 711 | |||
| XM_011520751.2 | XP_011519053.1 | 6 | 1137 | |||
| XM_011520752.2 | XP_011519054.1 | 6 | 1137 | |||
| XM_017019632.1 | XP_016875121.1 | 6 | 711 | |||
| XM_017019633.1 | XP_016875122.1 | 5 | 1103 | |||
| XM_017019634.1 | XP_016875123.1 | 5 | 1103 | |||
| XM_017019635.1 | XP_016875124.1 | 6 | 711 | |||
| XM_017019636.1 | XP_016875125.1 | 6 | 1137 | |||
| XM_017019637.1 | XP_016875126.1 | 3 | 270 | |||
| GenBank mRNA | AK001492.1 | - | 6 | 807 | ||
| AK001499.1 | - | 6 | 843 | |||
| AK225336.1 | - | 6 | 829 | |||
| AK297948.1 | - | 4 | 518 | |||
| BC003081.2 | - | 6 | 867 | 2989678 | ||
| BC008368.1 | - | 6 | 825 | 3355282 | ||

ASUN
Genomic Map