siRNA Details
| Gene Symbol: NUP133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018230.2 | NP_060700.2 | 11 | 1560 | ||
| GenBank mRNA | AK001676.1 | - | 11 | 1560 | ||
| AK001754.1 | - | 3 | 398 | |||
| AK092339.1 | - | 11 | 1517 | |||
| AK304061.1 | - | 12 | 1527 | |||
| AK304861.1 | - | 12 | 1461 | |||
| AK314431.1 | - | 11 | 1517 | |||
| BC020107.1 | - | 11 | 1544 | 3919973 | ||

NUP133
Genomic Map