siRNA Details
| Gene Symbol: AGK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018238.3 | NP_060708.1 | 6, 7 | 565 | ||
| XM_005250023.3 | XP_005250080.1 | 6, 7 | 461 | |||
| XM_011516397.2 | XP_011514699.1 | 6, 7 | 1031 | |||
| GenBank mRNA | AJ278150.1 | - | 5, 6 | 421 | ||
| AK001704.1 | - | 6, 7 | 433 | |||
| AK222769.1 | - | 6, 7 | 427 | |||
| AK299131.1 | - | 6, 7 | 424 | |||
| AK304502.1 | - | 7, 8 | 452 | |||
| BC022777.1 | - | 5, 6 | 419 | 3913448 | ||
| DB032353.1 | - | 6, 7 | 565 | |||

AGK
Genomic Map