siRNA Details
| Gene Symbol: TMEM30A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143958.1 | NP_001137430.1 | 5 | 906 | ||
| NM_018247.3 | NP_060717.1 | 6 | 1014 | |||
| GenBank mRNA | AK001718.1 | - | 6 | 883 | ||
| AK292823.1 | - | 5 | 771 | |||
| AK293225.1 | - | 7 | 839 | |||
| AK295567.1 | - | 7 | 835 | |||
| AL162046.1 | - | 3 | 242 | |||
| AL832490.1 | - | 6 | 937 | |||
| AL832815.1 | - | 6 | 876 | |||
| BC009006.1 | - | 5 | 757 | 4154200 | ||

TMEM30A
Genomic Map