siRNA Details
| Gene Symbol: TXLNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018360.2 | NP_060830.2 | 2 | 186 | ||
| GenBank mRNA | AK002071.1 | - | 2 | 181 | ||
| BC101572.1 | - | 2 | 153 | 8069078 | ||
| BC101576.1 | - | 2 | 153 | 8069082 | ||
| DB098819.1 | - | 2 | 186 | |||

TXLNG
Genomic Map