siRNA Details
| Gene Symbol: TXLNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168683.1 | NP_001162154.1 | 2 | 305 | ||
| NM_018360.2 | NP_060830.2 | 4 | 701 | |||
| GenBank mRNA | AK002071.1 | - | 4 | 696 | ||
| AY739713.1 | - | 2 | 255 | |||
| BC101572.1 | - | 4 | 668 | 8069078 | ||
| BC101576.1 | - | 4 | 668 | 8069082 | ||

TXLNG
Genomic Map