siRNA Details
| Gene Symbol: DOK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018431.4 | NP_060901.2 | 5 | 1047 | ||
| NM_177959.2 | NP_808874.1 | 5 | 693 | |||
| XM_011528904.1 | XP_011527206.1 | 5 | 676 | |||
| GenBank mRNA | AF132732.1 | - | 5 | 769 | ||
| AF466368.1 | - | 5 | 572 | |||
| AK313222.1 | - | 5 | 759 | |||
| AL050069.1 | - | 4 | 450 | |||
| BC008992.1 | - | 5 | 693 | 4183000 | ||

DOK5
Genomic Map