siRNA Details
Gene Symbol: RNF130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018434.5 | NP_060904.2 | 8 | 1643 | ||
GenBank mRNA | AF155650.1 | - | 7 | 942 | ||
AK313089.1 | - | 8 | 1126 | |||
AY083998.1 | - | 8 | 1625 | |||
BC017100.1 | - | 8 | 1225 | |||
BC065244.1 | - | 7 | 888 | |||
BC082267.1 | - | 8 | 1240 | |||
BC108306.1 | - | 8 | 1638 | 6137501 | ||
BC113864.1 | - | 9 | 1073 | 40033418 | ||
BC114472.1 | - | 8 | 1005 | |||
BQ447530.1 | - | 2 | 228 | |||
DB572650.2 | - | 1 | 69 |
