siRNA Details
| Gene Symbol: BRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018462.4 | NP_060932.2 | 1, 2 | 160 | ||
| GenBank mRNA | AF161418.1 | - | 1, 2 | 121 | ||
| AF220047.1 | - | 2, 3 | 1029 | |||
| AK312155.1 | - | 1, 2 | 134 | |||
| AY148219.1 | - | 1, 2 | 160 | |||
| AY148220.1 | - | 1, 2 | 160 | |||
| BC001067.1 | - | 1, 2 | 136 | 2823162 | ||
| BC007929.1 | - | 1, 2 | 129 | 3688264 | ||
| BC019303.1 | - | 1, 2 | 127 | 2823162 | ||
| BC061515.1 | - | 1, 2 | 134 | |||

BRK1
Genomic Map