siRNA Details
| Gene Symbol: USE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018467.3 | NP_060937.1 | 8 | 817 | ||
| XM_017026975.1 | XP_016882464.1 | 9 | 1527 | |||
| XM_017026976.1 | XP_016882465.1 | 7 | 767 | |||
| GenBank mRNA | AB097050.1 | - | 8 | 788 | ||
| AF220052.1 | - | 8 | 788 | |||
| BC006005.1 | - | 7 | 598 | 4279653 | ||
| BC008455.1 | - | 8 | 765 | 4247241 | ||
| BF448659.1 | - | 1 | 38 | |||
| T86451.1 | - | 1 | 37 | |||

USE1
Genomic Map