siRNA Details
| Gene Symbol: SLC22A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018484.3 | NP_060954.1 | 7, 8 | 1662 | ||
| XM_011545167.1 | XP_011543469.1 | 6, 7 | 1312 | |||
| GenBank mRNA | AB026116.1 | - | 7, 8 | 1315 | ||
| AK075224.1 | - | 7, 8 | 1008 | |||
| AK222869.1 | - | 7, 8 | 1320 | |||
| AK290791.1 | - | 7, 8 | 1645 | |||
| BC034384.1 | - | 7, 8 | 1637 | 5190509 | ||

SLC22A11
Genomic Map