siRNA Details
Gene Symbol: LMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001395.2 | NP_001001395.1 | 2 | 365 | ||
NM_001243609.1 | NP_001230538.1 | 3 | 432 | |||
NM_001243610.1 | NP_001230539.1 | 3 | 204 | |||
NM_001243611.1 | NP_001230540.1 | 2 | 601 | |||
NM_001243612.1 | NP_001230541.1 | 2 | 289 | |||
NM_001243613.1 | NP_001230542.1 | 3 | 303 | |||
NM_018640.4 | NP_061110.2 | 2 | 623 | |||
NR_045012.1 | 3 | 432 | ||||
NR_045013.1 | 3 | 432 | ||||
NR_045014.1 | 2 | 365 | ||||
XM_006719110.3 | XP_006719173.1 | 3 | 440 | |||
XM_006719111.2 | XP_006719174.1 | 2 | 483 | |||
XM_011520762.1 | XP_011519064.1 | 3 | 331 | |||
XM_011520763.1 | XP_011519065.1 | 3 | 238 | |||
XM_017019643.1 | XP_016875132.1 | 2 | 586 | |||
GenBank mRNA | AB044745.1 | - | 2 | 369 | ||
AB044746.1 | - | 4 | 270 | |||
AF258348.1 | - | 3 | 221 | |||
AK294474.1 | - | 2 | 289 | |||
AK294651.1 | - | 2 | 278 | |||
AK294909.1 | - | 3 | 303 | |||
AK308594.1 | - | 3 | 343 | |||
AK308826.1 | - | 4 | 308 | |||
AK308896.1 | - | 3 | 244 | |||
AK312418.1 | - | 2 | 507 | |||
AK316021.1 | - | 2 | 225 | |||
BC017777.1 | - | 2 | 178 | |||
BC026311.1 | - | 2 | 519 | 4799836 | ||
BC050085.1 | - | 3 | 205 | 5274811 | ||
BI553855.1 | - | 2 | 261 | |||
DA780743.1 | - | 2 | 366 |
