siRNA Details
| Gene Symbol: BIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018688.4 | NP_061158.1 | 2 | 137 | ||
| XM_005273573.3 | XP_005273630.1 | 2 | 105 | |||
| XM_011544586.2 | XP_011542888.1 | 2 | 109 | |||
| XM_011544587.1 | XP_011542889.1 | 2 | 137 | |||
| GenBank mRNA | AF271732.1 | - | 2 | 100 | ||
| AK001289.1 | - | 2 | 102 | |||
| AK023980.1 | - | 2 | 84 | |||
| BC001223.1 | - | 1 | 21 | 3534031 | ||
| BC009824.2 | - | 2 | 71 | 3953604 | ||
| DA480525.1 | - | 2 | 137 | |||

BIN3
Genomic Map