siRNA Details
| Gene Symbol: CTTNBP2NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018704.2 | NP_061174.1 | 3 | 258 | ||
| XM_011541781.2 | XP_011540083.1 | 3 | 329 | |||
| XM_017001806.1 | XP_016857295.1 | 3 | 696 | |||
| GenBank mRNA | AK022544.1 | - | 2 | 179 | ||
| AK023340.1 | - | 222 | ||||
| AL598321.1 | - | 3 | 214 | |||
| BC016029.1 | - | 3 | 198 | 3921313 | ||
| DA238082.1 | - | 3 | 187 | |||

CTTNBP2NL
Genomic Map