siRNA Details
| Gene Symbol: KLHL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018847.2 | NP_061335.1 | 1 | 1205 | ||
| GenBank mRNA | AB037775.1 | - | 1 | 1144 | ||
| AK091715.1 | - | 1 | 1155 | |||
| AL713669.1 | - | 1 | 1147 | |||
| AY172949.1 | - | 1 | 1164 | |||
| BC039133.2 | - | 1 | 1148 | 4339111 | ||
| BC105008.1 | - | 1 | 671 | 8144011 | ||
| BC113513.1 | - | 1 | 671 | 8322565 | ||
| BX538121.1 | - | 2 | 580 | |||

KLHL9
Genomic Map