siRNA Details
Gene Symbol: KLHL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018847.2 | NP_061335.1 | 1 | 616 | ||
GenBank mRNA | AB037775.1 | - | 1 | 555 | ||
AK091715.1 | - | 1 | 566 | |||
AL713669.1 | - | 1 | 558 | |||
AY172949.1 | - | 1 | 575 | |||
BC039133.2 | - | 1 | 559 | 4339111 | ||
BC105008.1 | - | 1 | 82 | 8144011 | ||
BC113513.1 | - | 1 | 82 | 8322565 |
