siRNA Details
| Gene Symbol: KLHL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031710.2 | NP_001026880.2 | 9 | 1503 | ||
| NM_018846.4 | NP_061334.4 | 9 | 1587 | |||
| NR_033328.1 | 10 | 1634 | ||||
| XM_006715753.2 | XP_006715816.1 | 10 | 1537 | |||
| XM_006715754.2 | XP_006715817.1 | 11 | 1620 | |||
| XM_006715755.2 | XP_006715818.1 | 11 | 1839 | |||
| XM_006715756.2 | XP_006715819.1 | 10 | 1708 | |||
| XM_017012439.1 | XP_016867928.1 | 10 | 1800 | |||
| GenBank mRNA | AK056390.1 | - | 10 | 1585 | ||
| AK290479.1 | - | 9 | 1297 | |||
| AK296219.1 | - | 6 | 1106 | |||
| AK297421.1 | - | 6 | 1027 | |||
| AK297595.1 | - | 8 | 1344 | |||
| AK299006.1 | - | 9 | 1229 | |||
| AL136597.1 | - | 9 | 1449 | |||
| AY028802.1 | - | 9 | 1152 | |||
| BC039585.1 | - | 9 | 1445 | 5527346 | ||
| EF560731.1 | - | 10 | 1353 | |||

KLHL7
Genomic Map