siRNA Details
| Gene Symbol: PDGFC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016205.2 | NP_057289.1 | 5 | 1310 | ||
| NR_036641.1 | 6 | 1371 | ||||
| XM_011532124.2 | XP_011530426.1 | 7 | 990 | |||
| XM_017008455.1 | XP_016863944.1 | 5 | 939 | |||
| GenBank mRNA | AB033831.1 | - | 5 | 1145 | ||
| AF091434.1 | - | 5 | 1310 | |||
| AF244813.1 | - | 5 | 1007 | |||
| AF260738.1 | - | 5 | 981 | |||
| AK300480.1 | - | 4 | 796 | |||
| AK304638.1 | - | 7 | 653 | |||
| AK313817.1 | - | 5 | 1040 | |||
| AM922296.1 | - | 6 | 1191 | |||
| AY358493.1 | - | 5 | 1103 | |||
| BC094746.1 | - | 5 | 911 | |||
| BC136662.1 | - | 5 | 1231 | 9020652 | ||

PDGFC
Genomic Map