siRNA Details
| Gene Symbol: PCDHGC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018929.2 | NP_061752.1 | 1 | 2063 | ||
| NM_032407.1 | NP_115783.1 | 1 | 2063 | |||
| GenBank mRNA | AF152339.1 | - | 1 | 2063 | ||
| AF152526.1 | - | 1 | 2063 | |||
| BC112081.1 | - | 1 | 2137 | 8327549 | ||
| BC112083.1 | - | 1 | 2137 | 8327551 | ||

PCDHGC5
Genomic Map