siRNA Details
| Gene Symbol: PCDHGB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018925.2 | NP_061748.1 | 1 | 1978 | ||
| NM_032099.1 | NP_115270.1 | 1 | 1978 | |||
| GenBank mRNA | AF152334.1 | - | 1 | 1978 | ||
| AF152521.1 | - | 1 | 1978 | |||
| BC093877.1 | - | 1 | 2069 | 7939722 | ||
| BC093879.1 | - | 1 | 2069 | 7939724 | ||

PCDHGB5
Genomic Map