siRNA Details
| Gene Symbol: PCDHGB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018924.4 | NP_061747.2 | 1 | 1007 | ||
| NM_032097.2 | NP_115268.2 | 1 | 1007 | |||
| GenBank mRNA | AF152332.1 | - | 1 | 876 | ||
| AF152519.1 | - | 1 | 876 | |||
| BC150172.1 | - | 1 | 1007 | 40147143 | ||

PCDHGB3
Genomic Map